Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTTTGCGACCAACCAGAATGCC[A/C]TAAAATTCACCCAAAGTGTACTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612173 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPAG16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPAG16 - sperm associated antigen 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VWC2L - von Willebrand factor C domain containing protein 2-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080500.2 | 402 | Missense Mutation | CAT,CCT | H,P 91 | NP_001073969.1 |