Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCTCCTCTGTAATAACTGGAGG[C/T]GATTGTTCGAAATCTTTCCTGGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616889 MIM: 179508 | ||||||||||||||||||||
Literature Links: |
CEP68 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEP68 - centrosomal protein 68 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB1A - RAB1A, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004161.4 | 615 | Missense Mutation | ACC,GCC | T,A 77 | NP_004152.1 | |
NM_015543.1 | 615 | Intron | NP_056358.1 | |||
XM_005264468.1 | 615 | Intron | XP_005264525.1 |