Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCTCAGGTCCAGTGTGGGGAGC[A/G]GCAGGAGCAGGAGGGCTGGGATCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605939 MIM: 604083 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLCD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLCD4 - phospholipase C delta 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF142 - zinc finger protein 142 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105537.2 | 6245 | Silent Mutation | GCC,GCT | A,A 1679 | NP_001099007.1 | |
XM_006712730.3 | 6245 | Silent Mutation | GCC,GCT | A,A 1879 | XP_006712793.1 | |
XM_011511784.2 | 6245 | Silent Mutation | GCC,GCT | A,A 1879 | XP_011510086.1 | |
XM_011511785.2 | 6245 | Silent Mutation | GCC,GCT | A,A 1879 | XP_011510087.1 | |
XM_011511786.2 | 6245 | Silent Mutation | GCC,GCT | A,A 1879 | XP_011510088.1 | |
XM_011511787.2 | 6245 | Silent Mutation | GCC,GCT | A,A 1879 | XP_011510089.1 | |
XM_011511788.2 | 6245 | Silent Mutation | GCC,GCT | A,A 1879 | XP_011510090.1 | |
XM_011511789.2 | 6245 | Silent Mutation | GCC,GCT | A,A 1679 | XP_011510091.1 | |
XM_017004871.1 | 6245 | Silent Mutation | GCC,GCT | A,A 1879 | XP_016860360.1 | |
XM_017004872.1 | 6245 | Silent Mutation | GCC,GCT | A,A 1679 | XP_016860361.1 |