Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCAGAACGCCTACAGCAAGTGCA[A/G]GGAATACAGTGACGACAAAGTGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608525 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ING5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ING5 - inhibitor of growth family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005247047.2 | 1320 | Missense Mutation | AAG,AGG | K,R 76 | XP_005247104.1 | |
XM_017005097.1 | 1320 | Missense Mutation | AAG,AGG | K,R 136 | XP_016860586.1 | |
XM_017005098.1 | 1320 | Missense Mutation | AAG,AGG | K,R 136 | XP_016860587.1 | |
XM_017005099.1 | 1320 | Missense Mutation | AAG,AGG | K,R 136 | XP_016860588.1 | |
XM_017005100.1 | 1320 | Missense Mutation | AAG,AGG | K,R 78 | XP_016860589.1 | |
XM_017005101.1 | 1320 | Missense Mutation | AAG,AGG | K,R 78 | XP_016860590.1 | |
XM_017005102.1 | 1320 | Missense Mutation | AAG,AGG | K,R 76 | XP_016860591.1 |