Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGGAGAAGGTGGAATCTTTGTCC[A/G]GTGATGGGGAAGGGGTAGGAGGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615900 | ||||||||||||||||||||
Literature Links: |
AGBL5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AGBL5 - ATP/GTP binding protein like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035507.2 | 266 | Missense Mutation | AGT,GGT | S,G 30 | NP_001030584.1 | |
NM_021831.5 | 266 | Missense Mutation | AGT,GGT | S,G 30 | NP_068603.4 | |
XM_005264477.2 | 266 | Missense Mutation | AGT,GGT | S,G 30 | XP_005264534.1 | |
XM_011533011.2 | 266 | Missense Mutation | AGT,GGT | S,G 30 | XP_011531313.1 | |
XM_011533012.2 | 266 | Missense Mutation | AGT,GGT | S,G 30 | XP_011531314.1 | |
XM_011533013.2 | 266 | Missense Mutation | AGT,GGT | S,G 30 | XP_011531315.1 |
AGBL5-AS1 - AGBL5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRA-AGC8-1 - transfer RNA-Ala (AGC) 8-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |