Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGAGCAATAGCAATGCGTTGTTT[C/G]TCCCCTCTAGAGAGTTGAGACCCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603201 | ||||||||||||||||||||
Literature Links: |
ABCB11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCB11 - ATP binding cassette subfamily B member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003742.2 | 2651 | Missense Mutation | GAC,GAG | D,E 1223 | NP_003733.2 | |
XM_006712817.3 | 2651 | Missense Mutation | GAC,GAG | D,E 1237 | XP_006712880.1 | |
XM_011512077.2 | 2651 | Missense Mutation | GAC,GAG | D,E 1257 | XP_011510379.1 | |
XM_011512078.2 | 2651 | Missense Mutation | GAC,GAG | D,E 1257 | XP_011510380.1 | |
XM_011512080.2 | 2651 | Intron | XP_011510382.1 | |||
XM_011512081.2 | 2651 | Missense Mutation | GAC,GAG | D,E 665 | XP_011510383.1 | |
XM_017005165.1 | 2651 | Missense Mutation | GAC,GAG | D,E 1257 | XP_016860654.1 | |
XM_017005166.1 | 2651 | Missense Mutation | GAC,GAG | D,E 1000 | XP_016860655.1 | |
XM_017005167.1 | 2651 | Missense Mutation | GAC,GAG | D,E 818 | XP_016860656.1 |