Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCAGTTCCTGGAACTGCTGAGG[A/T]CCCACCAGAGCCGACCAGCAAAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605788 MIM: 615301 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAPRE3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAPRE3 - microtubule associated protein RP/EB family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM214 - transmembrane protein 214 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083590.1 | 822 | Missense Mutation | ACC,TCC | T,S 202 | NP_001077059.1 | |
NM_017727.4 | 822 | Missense Mutation | ACC,TCC | T,S 247 | NP_060197.4 | |
XM_005264381.3 | 822 | Missense Mutation | ACC,TCC | T,S 247 | XP_005264438.1 | |
XM_005264382.3 | 822 | Missense Mutation | ACC,TCC | T,S 247 | XP_005264439.1 | |
XM_005264383.3 | 822 | Intron | XP_005264440.1 |