Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGTTGTGTGGAATGAGGTACAG[C/T]TCTCTGAACGCAAGAACTACAAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606010 | ||||||||||||||||||||
Literature Links: |
KRTCAP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KRTCAP3 - keratinocyte associated protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NRBP1 - nuclear receptor binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321357.1 | 1133 | Missense Mutation | CTC,TTC | L,F 101 | NP_001308286.1 | |
NM_001321358.1 | 1133 | Missense Mutation | CTC,TTC | L,F 101 | NP_001308287.1 | |
NM_001321359.1 | 1133 | Missense Mutation | CTC,TTC | L,F 101 | NP_001308288.1 | |
NM_001321361.1 | 1133 | Missense Mutation | CTC,TTC | L,F 101 | NP_001308290.1 | |
NM_001321362.1 | 1133 | Missense Mutation | CTC,TTC | L,F 101 | NP_001308291.1 | |
NM_001321363.1 | 1133 | Missense Mutation | CTC,TTC | L,F 101 | NP_001308292.1 | |
NM_013392.3 | 1133 | Missense Mutation | CTC,TTC | L,F 101 | NP_037524.1 |