Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCAGGGGTCTTGCTGGTGGCTG[A/C]CAGGCCCCCCAGGCTCTGAAGGCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf50 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2orf50 - chromosome 2 open reading frame 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182500.2 | 420 | Nonsense Mutation | TGA,TGC | *,C 46 | NP_872306.1 | |
XM_006711857.2 | 420 | Nonsense Mutation | TGA,TGC | *,C 46 | XP_006711920.1 | |
XM_006711858.2 | 420 | Nonsense Mutation | TGA,TGC | *,C 46 | XP_006711921.1 | |
XM_006711859.3 | 420 | Nonsense Mutation | TGA,TGC | *,C 46 | XP_006711922.1 |
FLJ33534 - uncharacterized LOC285150 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |