Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTATATCACAGCTTCACCACGGTC[A/G]GAGGCAAGGCCGACTTCAATTAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614967 | ||||||||||||||||||||
Literature Links: |
GPR155 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR155 - G protein-coupled receptor 155 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033045.3 | 2829 | Silent Mutation | NP_001028217.1 | |||
NM_001267050.1 | 2829 | Silent Mutation | NP_001253979.1 | |||
NM_001267051.1 | 2829 | Silent Mutation | NP_001253980.1 | |||
NM_152529.6 | 2829 | Silent Mutation | NP_689742.4 | |||
XM_017003485.1 | 2829 | Silent Mutation | XP_016858974.1 | |||
XM_017003486.1 | 2829 | Silent Mutation | XP_016858975.1 | |||
XM_017003487.1 | 2829 | Silent Mutation | XP_016858976.1 |
SCRN3 - secernin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |