Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTGACGGTGATCCAGGGTGGCGC[A/T]GGCTGCCCATCAGCTCCAGGCCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1211L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA1211L - KIAA1211 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207362.2 | 2760 | Silent Mutation | CCA,CCT | P,P 822 | NP_997245.2 | |
XM_005263923.3 | 2760 | Silent Mutation | CCA,CCT | P,P 850 | XP_005263980.1 | |
XM_005263924.3 | 2760 | Silent Mutation | CCA,CCT | P,P 850 | XP_005263981.1 | |
XM_005263926.3 | 2760 | Silent Mutation | CCA,CCT | P,P 822 | XP_005263983.1 | |
XM_011511093.1 | 2760 | Silent Mutation | CCA,CCT | P,P 836 | XP_011509395.1 | |
XM_011511094.1 | 2760 | Silent Mutation | CCA,CCT | P,P 822 | XP_011509396.1 | |
XM_011511095.2 | 2760 | Silent Mutation | CCA,CCT | P,P 822 | XP_011509397.1 | |
XM_011511096.1 | 2760 | Silent Mutation | CCA,CCT | P,P 689 | XP_011509398.1 | |
XM_017003971.1 | 2760 | Silent Mutation | CCA,CCT | P,P 836 | XP_016859460.1 | |
XM_017003972.1 | 2760 | Silent Mutation | CCA,CCT | P,P 836 | XP_016859461.1 | |
XM_017003973.1 | 2760 | Intron | XP_016859462.1 |