Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACACCACAATCCGTCAAAAGCGA[C/T]TTTCTTCCTCTTCCACAGCACGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604448 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYTIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYTIP - cytohesin 1 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004288.4 | 1333 | Missense Mutation | AAT,AGT | N,S 357 | NP_004279.3 | |
XM_017005386.1 | 1333 | Missense Mutation | AAT,AGT | N,S 251 | XP_016860875.1 |