Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAACAGCGTATCCCGCACAGGTT[C/T]GAACCCCACATTGCGGGGGGTACCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602878 | ||||||||||||||||||||
Literature Links: |
SLC30A3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC30A3 - solute carrier family 30 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318949.1 | 950 | Missense Mutation | AAA,GAA | K,E 298 | NP_001305878.1 | |
NM_001318950.1 | 950 | Missense Mutation | AAA,GAA | K,E 290 | NP_001305879.1 | |
NM_001318951.1 | 950 | Missense Mutation | AAA,GAA | K,E 290 | NP_001305880.1 | |
NM_003459.4 | 950 | Missense Mutation | AAA,GAA | K,E 303 | NP_003450.2 | |
XM_005264547.1 | 950 | Silent Mutation | TCA,TCG | S,S 251 | XP_005264604.1 | |
XM_005264548.3 | 950 | Silent Mutation | TCA,TCG | S,S 246 | XP_005264605.1 | |
XM_006712100.2 | 950 | Silent Mutation | TCA,TCG | S,S 202 | XP_006712163.1 | |
XM_011533102.2 | 950 | Missense Mutation | AAA,GAA | K,E 230 | XP_011531404.1 | |
XM_011533103.2 | 950 | Missense Mutation | AAA,GAA | K,E 189 | XP_011531405.1 | |
XM_017004873.1 | 950 | Silent Mutation | TCA,TCG | S,S 238 | XP_016860362.1 | |
XM_017004874.1 | 950 | Silent Mutation | TCA,TCG | S,S 238 | XP_016860363.1 | |
XM_017004875.1 | 950 | Silent Mutation | TCA,TCG | S,S 229 | XP_016860364.1 |