Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGGCCCCGGCATGGCGGGGCTGG[C/G]CAGCCATCACAACCAGGGCCTGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604462 | ||||||||||||||||||||
Literature Links: |
ANKRD39 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD39 - ankyrin repeat domain 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEMA4C - semaphorin 4C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017789.4 | 2010 | Missense Mutation | GCC,GGC | A,G 617 | NP_060259.4 | |
XM_006712606.3 | 2010 | Intron | XP_006712669.2 | |||
XM_011511378.2 | 2010 | Missense Mutation | GCC,GGC | A,G 617 | XP_011509680.1 | |
XM_011511379.2 | 2010 | Missense Mutation | GCC,GGC | A,G 617 | XP_011509681.1 | |
XM_011511380.1 | 2010 | Missense Mutation | GCC,GGC | A,G 617 | XP_011509682.1 | |
XM_011511381.1 | 2010 | Missense Mutation | GCC,GGC | A,G 617 | XP_011509683.1 | |
XM_011511382.2 | 2010 | Missense Mutation | GCC,GGC | A,G 617 | XP_011509684.1 | |
XM_011511383.1 | 2010 | Missense Mutation | GCC,GGC | A,G 617 | XP_011509685.1 | |
XM_017004393.1 | 2010 | Missense Mutation | GCC,GGC | A,G 659 | XP_016859882.1 | |
XM_017004394.1 | 2010 | Missense Mutation | GCC,GGC | A,G 476 | XP_016859883.1 |