Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAACTTGCAGACATAGGGTAACC[A/T]CACATTACAGTTATAATCTTTCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 167805 | ||||||||||||||||||||
Literature Links: |
REG3A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
REG3A - regenerating family member 3 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002580.2 | 557 | Missense Mutation | AGG,TGG | R,W 166 | NP_002571.1 | |
NM_138937.2 | 557 | Missense Mutation | AGG,TGG | R,W 166 | NP_620354.1 | |
NM_138938.2 | 557 | Intron | NP_620355.1 |