Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCTGGATTCATCTTCAGACACC[C/G]CTTTGGAGGAAGAGAATAGCAAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CDKL4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDKL4 - cyclin dependent kinase like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009565.1 | 2209 | Missense Mutation | CGG,GGG | R,G 265 | NP_001009565.1 | |
XM_011532815.2 | 2209 | Missense Mutation | CGG,GGG | R,G 265 | XP_011531117.1 | |
XM_011532816.2 | 2209 | Missense Mutation | CGG,GGG | R,G 265 | XP_011531118.1 | |
XM_011532817.2 | 2209 | Missense Mutation | CGG,GGG | R,G 224 | XP_011531119.1 | |
XM_011532819.2 | 2209 | Intron | XP_011531121.1 | |||
XM_017003981.1 | 2209 | Missense Mutation | CGG,GGG | R,G 265 | XP_016859470.1 | |
XM_017003982.1 | 2209 | Missense Mutation | CGG,GGG | R,G 265 | XP_016859471.1 | |
XM_017003983.1 | 2209 | Intron | XP_016859472.1 | |||
XM_017003984.1 | 2209 | Intron | XP_016859473.1 | |||
XM_017003985.1 | 2209 | Intron | XP_016859474.1 |