Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTTTCTCTCCTTGTTTTGTGCAG[A/T]CGGCGGAGACTGGCTGAGTGGAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611678 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDCL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDCL3 - phosducin like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024065.4 | 466 | Intron | NP_076970.1 | |||
XM_017004890.1 | 466 | Missense Mutation | AGA,AGT | R,S 25 | XP_016860379.1 |