Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGGCGGCAGCGCTGGCTGCAGCG[A/C]GGCCTGGGGCAGCGCGGGCGGCGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601135 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GBX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GBX2 - gastrulation brain homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301687.1 | 243 | Missense Mutation | GCG,TCG | A,S 69 | NP_001288616.1 | |
NM_001485.3 | 243 | Missense Mutation | GCG,TCG | A,S 69 | NP_001476.2 |
LOC105373944 - translation initiation factor IF-2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |