Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGTCTCCTATTTCCAGAACGCCA[A/G]TCTGAATTGCCAACGCAAAGCAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CNOT11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNOT11 - CCR4-NOT transcription complex subunit 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017546.4 | 769 | Silent Mutation | CAA,CAG | Q,Q 229 | NP_060016.3 |
RNF149 - ring finger protein 149 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |