Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCTGCTTGGCTCTGCACTCTAC[C/G]ATAAAGCTGTTTCTCACCAGGAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604107 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR55 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR55 - G protein-coupled receptor 55 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005683.3 | 964 | Silent Mutation | ATC,ATG | I,M 257 | NP_005674.2 | |
XM_005246952.3 | 964 | Missense Mutation | ATC,ATG | I,M 257 | XP_005247009.1 | |
XM_011512175.2 | 964 | Silent Mutation | ATC,ATG | I,M 257 | XP_011510477.1 | |
XM_011512176.2 | 964 | Silent Mutation | ATC,ATG | I,M 257 | XP_011510478.1 |
LOC151484 - uncharacterized LOC151484 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |