Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGCAAACAGCCACGGGTCAGCC[C/G]AGGACCAGTCCACGGCATAGACGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608998 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSSC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TSSC1 - tumor suppressing subtransferable candidate 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_006711893.2 | 1168 | Missense Mutation | TCG,TGG | S,W 257 | XP_006711956.1 | |
XM_011510383.2 | 1168 | Missense Mutation | TCG,TGG | S,W 396 | XP_011508685.1 | |
XM_011510384.2 | 1168 | Missense Mutation | TCG,TGG | S,W 384 | XP_011508686.1 | |
XM_011510385.2 | 1168 | Missense Mutation | TCG,TGG | S,W 369 | XP_011508687.1 | |
XM_011510387.2 | 1168 | Missense Mutation | TCG,TGG | S,W 213 | XP_011508689.1 | |
XM_017004818.1 | 1168 | Missense Mutation | TCG,TGG | S,W 257 | XP_016860307.1 |