Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGATGCCGTCCGTGCAGCGGCTG[G/T]AGGACTTCCACAGCGGCCGCAGCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604787 | ||||||||||||||||||||
Literature Links: |
ARL4C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARL4C - ADP ribosylation factor like GTPase 4C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282431.1 | 712 | Missense Mutation | TAC,TCC | Y,S 82 | NP_001269360.1 | |
NM_005737.3 | 712 | Missense Mutation | TAC,TCC | Y,S 82 | NP_005728.2 |