Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAAAGAAAAAAAAATCATACCTA[C/T]ATCCAAATATGTTATATTAAAACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603424 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927027 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101927027 - uncharacterized LOC101927027 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRKRA - protein activator of interferon induced protein kinase EIF2AK2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139517.1 | 1023 | Missense Mutation | ATA,GTA | I,V 250 | NP_001132989.1 | |
NM_001139518.1 | 1023 | Missense Mutation | ATA,GTA | I,V 236 | NP_001132990.1 | |
NM_001316362.1 | 1023 | Missense Mutation | ATA,GTA | I,V 148 | NP_001303291.1 | |
NM_003690.4 | 1023 | Missense Mutation | ATA,GTA | I,V 261 | NP_003681.1 | |
XM_011512063.1 | 1023 | Missense Mutation | ATA,GTA | I,V 176 | XP_011510365.1 | |
XM_011512066.1 | 1023 | Missense Mutation | ATA,GTA | I,V 148 | XP_011510368.1 | |
XM_017005159.1 | 1023 | Missense Mutation | ATA,GTA | I,V 148 | XP_016860648.1 |