Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCTCACCTGCTCAAATTCAGACT[C/T]CAATTCCTGTTCTTGTACCCTAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600378 MIM: 614918 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IMMT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IMMT - inner membrane mitochondrial protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100169.1 | 1901 | Missense Mutation | AAG,GAG | K,E 505 | NP_001093639.1 | |
NM_001100170.1 | 1901 | Missense Mutation | AAG,GAG | K,E 495 | NP_001093640.1 | |
NM_006839.2 | 1901 | Missense Mutation | AAG,GAG | K,E 506 | NP_006830.2 | |
XM_005264110.3 | 1901 | Intron | XP_005264167.1 | |||
XM_005264113.1 | 1901 | Intron | XP_005264170.1 | |||
XM_005264114.1 | 1901 | Intron | XP_005264171.1 | |||
XM_011532497.2 | 1901 | Intron | XP_011530799.1 | |||
XM_017003194.1 | 1901 | Intron | XP_016858683.1 | |||
XM_017003195.1 | 1901 | Intron | XP_016858684.1 |
PTCD3 - pentatricopeptide repeat domain 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |