Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGACAAGCCGTGGATGGTAGATC[A/T]TCATGGGTCTGAGGAGAGAGAATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 194500 | ||||||||||||||||||||
Literature Links: |
ZNF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF2 - zinc finger protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017396.2 | 713 | Missense Mutation | CAT,CTT | H,L 35 | NP_001017396.1 | |
NM_001282398.1 | 713 | Intron | NP_001269327.1 | |||
NM_001291604.1 | 713 | Intron | NP_001278533.1 | |||
NM_001291605.1 | 713 | Missense Mutation | CAT,CTT | H,L 77 | NP_001278534.1 | |
NM_021088.3 | 713 | Missense Mutation | CAT,CTT | H,L 77 | NP_066574.2 |