Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGGGCAGGGGTCGTCTGCCCCAC[C/T]CAACGGCGGGTCGAATTCGCACCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602919 MIM: 607163 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DOK1 - docking protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197260.1 | 588 | Intron | NP_001184189.1 | |||
NM_001318866.1 | 588 | Intron | NP_001305795.1 | |||
NM_001318867.1 | 588 | Intron | NP_001305796.1 | |||
NM_001318868.1 | 588 | Intron | NP_001305797.1 | |||
NM_001318869.1 | 588 | Intron | NP_001305798.1 | |||
NM_001381.4 | 588 | Intron | NP_001372.1 | |||
XM_017003528.1 | 588 | Intron | XP_016859017.1 |
LOXL3 - lysyl oxidase like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289164.1 | 588 | Intron | NP_001276093.1 | |||
NM_001289165.1 | 588 | UTR 5 | NP_001276094.1 | |||
NM_032603.3 | 588 | Missense Mutation | NP_115992.1 | |||
XM_011533134.2 | 588 | Missense Mutation | XP_011531436.1 | |||
XM_017005112.1 | 588 | UTR 5 | XP_016860601.1 |
M1AP - meiosis 1 associated protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |