Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTCTCCCGGCTGCTCCTCGCCAC[C/T]GGCCACTGGAAGAGGCTGCGGCGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608291 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TTL - tubulin tyrosine ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153712.4 | 290 | Silent Mutation | ACC,ACT | T,T 24 | NP_714923.1 | |
XM_005263599.3 | 290 | Silent Mutation | ACC,ACT | T,T 24 | XP_005263656.1 | |
XM_011510665.2 | 290 | Silent Mutation | ACC,ACT | T,T 24 | XP_011508967.1 |