Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGATGGAGAAGCAGCGGGCACTC[A/G]TGGCCGCCAAGGATGGGGATGTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611056 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESPNL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ESPNL - espin-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308370.1 | 132 | Intron | NP_001295299.1 | |||
NM_194312.3 | 132 | Missense Mutation | ATG,GTG | M,V 8 | NP_919288.2 | |
XM_011511087.1 | 132 | Intron | XP_011509389.1 |
SCLY - selenocysteine lyase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBE2F-SCLY - UBE2F-SCLY readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |