Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTCCTTCCTCTGTGTAGGAGTTG[A/G]TGTCCTGCCTGCGCTCAGGATGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612502 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COLEC11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COLEC11 - collectin subfamily member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001255982.1 | 344 | UTR 5 | NP_001242911.1 | |||
NM_001255983.1 | 344 | UTR 5 | NP_001242912.1 | |||
NM_001255984.1 | 344 | UTR 5 | NP_001242913.1 | |||
NM_001255985.1 | 344 | Missense Mutation | GAT,GGT | D,G 8 | NP_001242914.1 | |
NM_001255986.1 | 344 | Intron | NP_001242915.1 | |||
NM_001255987.1 | 344 | Intron | NP_001242916.1 | |||
NM_001255988.1 | 344 | Intron | NP_001242917.1 | |||
NM_001255989.1 | 344 | Intron | NP_001242918.1 | |||
NM_024027.4 | 344 | UTR 5 | NP_076932.1 | |||
NM_199235.2 | 344 | UTR 5 | NP_954705.1 | |||
XM_005263853.4 | 344 | Missense Mutation | GAT,GGT | D,G 8 | XP_005263910.1 | |
XM_006711897.3 | 344 | Missense Mutation | GAT,GGT | D,G 8 | XP_006711960.1 |