Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGAGGACTAGCTCGGGCACCTCC[A/G]TTGCTGGCAACCCATTCCAGGCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606622 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMARCAL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMARCAL1 - SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127207.1 | 268 | Missense Mutation | ATT,GTT | I,V 42 | NP_001120679.1 | |
NM_014140.3 | 268 | Missense Mutation | ATT,GTT | I,V 42 | NP_054859.2 | |
XM_005246631.2 | 268 | Missense Mutation | ATT,GTT | I,V 42 | XP_005246688.1 | |
XM_005246632.1 | 268 | Missense Mutation | ATT,GTT | I,V 42 | XP_005246689.1 | |
XM_017004228.1 | 268 | UTR 5 | XP_016859717.1 |