Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGTAATTGTGCTGTGAGAGCAGT[C/T]GTACTATTATCTGGTGCCTCGCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605295 | ||||||||||||||||||||
Literature Links: |
FIGN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FIGN - fidgetin, microtubule severing factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321825.1 | 2306 | Missense Mutation | CAA,CGA | Q,R 653 | NP_001308754.1 | |
NM_018086.3 | 2306 | Missense Mutation | CAA,CGA | Q,R 664 | NP_060556.2 | |
XM_011511389.2 | 2306 | Missense Mutation | CAA,CGA | Q,R 690 | XP_011509691.1 | |
XM_017004419.1 | 2306 | Missense Mutation | CAA,CGA | Q,R 690 | XP_016859908.1 |