Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGACCGGGCTGGGGCAGACATGTG[A/G]GCCAGCACCTTCACCCTGGCCATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RTP5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RTP5 - receptor transporter protein 5 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173821.2 | 50 | Nonsense Mutation | TGA,TGG | *,W 9 | NP_776182.2 | |
XM_011510995.2 | 50 | Intron | XP_011509297.1 | |||
XM_011510997.2 | 50 | Nonsense Mutation | TGA,TGG | *,W 9 | XP_011509299.1 |