Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACCTGCCCAGGCCAAACGCCTGG[A/C]ACAGCCAAAGTCAATCAGCTTTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP3K19 - mitogen-activated protein kinase kinase kinase 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018044.2 | 3314 | Missense Mutation | GCC,TCC | A,S 1095 | NP_001018054.1 | |
NM_001018046.2 | 3314 | Missense Mutation | GCC,TCC | A,S 390 | NP_001018056.1 | |
NM_001018047.2 | 3314 | Missense Mutation | GCC,TCC | A,S 342 | NP_001018057.1 | |
NM_001282883.1 | 3314 | Missense Mutation | GCC,TCC | A,S 340 | NP_001269812.1 | |
NM_001321177.1 | 3314 | UTR 3 | NP_001308106.1 | |||
NM_025052.4 | 3314 | Missense Mutation | GCC,TCC | A,S 1208 | NP_079328.3 | |
XM_011511891.2 | 3314 | Missense Mutation | GCC,TCC | A,S 1225 | XP_011510193.1 | |
XM_011511892.1 | 3314 | Missense Mutation | GCC,TCC | A,S 1208 | XP_011510194.1 | |
XM_011511894.2 | 3314 | Missense Mutation | GCC,TCC | A,S 1180 | XP_011510196.1 | |
XM_011511896.2 | 3314 | Missense Mutation | GCC,TCC | A,S 1113 | XP_011510198.1 | |
XM_011511897.2 | 3314 | Missense Mutation | GCC,TCC | A,S 1113 | XP_011510199.1 | |
XM_017005003.1 | 3314 | Missense Mutation | GCC,TCC | A,S 1208 | XP_016860492.1 | |
XM_017005004.1 | 3314 | Missense Mutation | GCC,TCC | A,S 1180 | XP_016860493.1 | |
XM_017005005.1 | 3314 | Missense Mutation | GCC,TCC | A,S 1180 | XP_016860494.1 | |
XM_017005006.1 | 3314 | Missense Mutation | GCC,TCC | A,S 1086 | XP_016860495.1 | |
XM_017005007.1 | 3314 | UTR 3 | XP_016860496.1 |