Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTATTTGGTCATCGACCTTACTG[A/G]TGGGTCCAAGAAACTCAGATTTACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608058 | ||||||||||||||||||||
Literature Links: |
G6PC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
G6PC2 - glucose-6-phosphatase catalytic subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081686.1 | 338 | Nonsense Mutation | TGA,TGG | *,W 82 | NP_001075155.1 | |
NM_021176.2 | 338 | Nonsense Mutation | TGA,TGG | *,W 82 | NP_066999.1 | |
XM_011511564.2 | 338 | Nonsense Mutation | TGA,TGG | *,W 82 | XP_011509866.1 | |
XM_011511565.2 | 338 | Intron | XP_011509867.1 |