Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAACCCGGCCCCCGAAGGGCCGCC[C/G]TCCGGGAGACGGTGATGCTGTTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 192975 | ||||||||||||||||||||
Literature Links: |
ITGA4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ITGA4 - integrin subunit alpha 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000885.5 | 809 | Missense Mutation | CTC,GTC | L,V 16 | NP_000876.3 | |
NM_001316312.1 | 809 | Missense Mutation | CTC,GTC | L,V 16 | NP_001303241.1 |