Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTAAACCCAGGACGATTTATTCCA[A/G]TTTGCAGTTGCAGGCTTTGAACCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600029 MIM: 610267 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DLX1 - distal-less homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001038493.1 | 608 | Intron | NP_001033582.1 | |||
NM_178120.4 | 608 | Missense Mutation | AAT,AGT | N,S 137 | NP_835221.2 |
METAP1D - methionyl aminopeptidase type 1D (mitochondrial) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |