Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAGGGGAGCGGTACCTTTCTCG[A/G]AGCTCGGCTCTCCCAGTGTGGGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602919 MIM: 607163 | ||||||||||||||||||||
Literature Links: |
DOK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DOK1 - docking protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOXL3 - lysyl oxidase like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
M1AP - meiosis 1 associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281295.1 | 700 | Intron | NP_001268224.1 | |||
NM_001281296.1 | 700 | Missense Mutation | CCG,TCG | P,S 472 | NP_001268225.1 | |
NM_001321739.1 | 700 | Missense Mutation | CCG,TCG | P,S 472 | NP_001308668.1 | |
NM_138804.4 | 700 | Missense Mutation | CCG,TCG | P,S 472 | NP_620159.2 | |
XM_005264152.3 | 700 | Missense Mutation | CCG,TCG | P,S 228 | XP_005264209.1 | |
XM_006711946.3 | 700 | Missense Mutation | CCG,TCG | P,S 510 | XP_006712009.1 | |
XM_011532548.2 | 700 | Missense Mutation | CCG,TCG | P,S 510 | XP_011530850.1 | |
XM_011532549.2 | 700 | Missense Mutation | CCG,TCG | P,S 490 | XP_011530851.1 | |
XM_011532550.2 | 700 | Missense Mutation | CCG,TCG | P,S 472 | XP_011530852.1 | |
XM_011532551.2 | 700 | Missense Mutation | CCG,TCG | P,S 472 | XP_011530853.1 | |
XM_011532552.2 | 700 | Missense Mutation | CCG,TCG | P,S 452 | XP_011530854.1 | |
XM_011532553.2 | 700 | Missense Mutation | CCG,TCG | P,S 441 | XP_011530855.1 | |
XM_011532554.2 | 700 | Missense Mutation | CCG,TCG | P,S 228 | XP_011530856.1 |