Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCCTTGGCAATTTCACTAAGCAG[C/G]TGGATGTAATCTGTATTTGGAATAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603424 | ||||||||||||||||||||
Literature Links: |
LOC101927027 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101927027 - uncharacterized LOC101927027 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRKRA - protein activator of interferon induced protein kinase EIF2AK2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139517.1 | 971 | Missense Mutation | CAC,CAG | H,Q 232 | NP_001132989.1 | |
NM_001139518.1 | 971 | Missense Mutation | CAC,CAG | H,Q 218 | NP_001132990.1 | |
NM_001316362.1 | 971 | Missense Mutation | CAC,CAG | H,Q 130 | NP_001303291.1 | |
NM_003690.4 | 971 | Missense Mutation | CAC,CAG | H,Q 243 | NP_003681.1 | |
XM_011512063.1 | 971 | Missense Mutation | CAC,CAG | H,Q 158 | XP_011510365.1 | |
XM_011512066.1 | 971 | Missense Mutation | CAC,CAG | H,Q 130 | XP_011510368.1 | |
XM_017005159.1 | 971 | Missense Mutation | CAC,CAG | H,Q 130 | XP_016860648.1 |