Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTCGATGAGGTGCTCCAGGCAGT[C/T]GGGGTGCCGGGTGCGCACTGCCACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610736 MIM: 607804 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD23 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD23 - ankyrin repeat domain 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144994.7 | 699 | Missense Mutation | AAC,GAC | N,D 224 | NP_659431.5 |
ANKRD39 - ankyrin repeat domain 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CNNM3 - cyclin and CBS domain divalent metal cation transport mediator 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |