Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTCTTTCCATCAACTGGGTCCCA[A/C]TGTAGAATTCCCAGGGGTTGGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602452 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BUB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BUB1 - BUB1 mitotic checkpoint serine/threonine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278616.1 | 2550 | Missense Mutation | AGT,ATT | S,I 813 | NP_001265545.1 | |
NM_001278617.1 | 2550 | Missense Mutation | AGT,ATT | S,I 833 | NP_001265546.1 | |
NM_004336.4 | 2550 | Missense Mutation | AGT,ATT | S,I 833 | NP_004327.1 |