Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTAAGGAAAACATAAATGGGCT[C/T]TCAGGATTCTTAGGGTCAATATTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607496 MIM: 609395 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOSTRIN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOSTRIN - nitric oxide synthase trafficking | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPC25 - SPC25, NDC80 kinetochore complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020675.3 | 641 | Silent Mutation | GAA,GAG | E,E 167 | NP_065726.1 | |
XM_011511516.2 | 641 | Silent Mutation | GAA,GAG | E,E 167 | XP_011509818.1 |