Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCATCCCCCCGTAGAGCCTGGC[C/G]CTCCTCGTCGCTTCTCTGCAGTAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602663 MIM: 602206 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRLH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRLH - prolactin releasing hormone | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB17 - RAB17, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022449.3 | 1108 | Missense Mutation | CGC,GGC | R,G 188 | NP_071894.1 | |
XM_006712689.1 | 1108 | Missense Mutation | CGC,GGC | R,G 166 | XP_006712752.1 | |
XM_017004693.1 | 1108 | Intron | XP_016860182.1 | |||
XM_017004694.1 | 1108 | Intron | XP_016860183.1 |