Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCACACTGCAACAGCTGAACCAC[A/G]TATTTGAGCTGCACCTGGGGCCATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607172 | ||||||||||||||||||||
Literature Links: |
STK11IP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
STK11IP - serine/threonine kinase 11 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052902.3 | 209 | Missense Mutation | ATA,GTA | I,V 44 | NP_443134.3 | |
XM_011510526.1 | 209 | Missense Mutation | ATA,GTA | I,V 44 | XP_011508828.1 | |
XM_011510527.2 | 209 | Missense Mutation | ATA,GTA | I,V 44 | XP_011508829.1 | |
XM_011510528.2 | 209 | Intron | XP_011508830.1 | |||
XM_011510529.1 | 209 | Missense Mutation | ATA,GTA | I,V 44 | XP_011508831.1 |