Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCAGCGGGAAGACCAGCAAGTCC[A/G]AACCGAACCATGTTATCTTCAAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 181031 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SAG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SAG - S-antigen visual arrestin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000541.4 | 108 | Missense Mutation | AAA,GAA | K,E 11 | NP_000532.2 | |
XM_011511593.2 | 108 | Intron | XP_011509895.1 | |||
XM_011511594.1 | 108 | Intron | XP_011509896.1 | |||
XM_011511596.1 | 108 | Intron | XP_011509898.1 | |||
XM_017004641.1 | 108 | Missense Mutation | AAA,GAA | K,E 11 | XP_016860130.1 | |
XM_017004642.1 | 108 | Missense Mutation | AAA,GAA | K,E 11 | XP_016860131.1 | |
XM_017004643.1 | 108 | Missense Mutation | AAA,GAA | K,E 11 | XP_016860132.1 |