Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGGTGGTGTCGCAGCAACACTT[A/T]CCTTTTGCTCGTTCTCCATCTAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600073 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRP2 - LDL receptor related protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004525.2 | 13905 | Missense Mutation | GAA,GTA | E,V 4607 | NP_004516.2 | |
XM_011511183.2 | 13905 | Missense Mutation | GAA,GTA | E,V 4564 | XP_011509485.1 | |
XM_011511184.2 | 13905 | Missense Mutation | GAA,GTA | E,V 3844 | XP_011509486.1 |