Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCACCAGTTTGTTAGAACCACGAA[A/C]AACAATTGTAACTGTTTTTCCAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605142 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCT4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCT4 - chaperonin containing TCP1 subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256721.1 | 1332 | Missense Mutation | GTT,TTT | V,F 360 | NP_001243650.1 | |
NM_006430.3 | 1332 | Missense Mutation | GTT,TTT | V,F 390 | NP_006421.2 |