Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAATGTCAGGATATAGGTACTTCA[A/G]AGGGTTTTCAGGAATGTTTTCAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600558 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105373805 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105373805 - uncharacterized LOC105373805 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STAT4 - signal transducer and activator of transcription 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243835.1 | 2287 | Silent Mutation | CTG,TTG | L,L 658 | NP_001230764.1 | |
NM_003151.3 | 2287 | Silent Mutation | CTG,TTG | L,L 658 | NP_003142.1 | |
XM_006712719.3 | 2287 | Silent Mutation | CTG,TTG | L,L 658 | XP_006712782.1 | |
XM_011511705.2 | 2287 | Silent Mutation | CTG,TTG | L,L 658 | XP_011510007.1 | |
XM_017004784.1 | 2287 | Intron | XP_016860273.1 |