Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGAGCAGTGGGGGTGGTTGTGGTT[A/G]TGTGCAGGACCTTAGCTCGGACCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611437 | ||||||||||||||||||||
Literature Links: |
DUSP19 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DUSP19 - dual specificity phosphatase 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142314.1 | 542 | Missense Mutation | TAT,TGT | Y,C 56 | NP_001135786.1 | |
NM_001321519.1 | 542 | Missense Mutation | TAT,TGT | Y,C 56 | NP_001308448.1 | |
NM_080876.3 | 542 | Missense Mutation | TAT,TGT | Y,C 56 | NP_543152.1 |