Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCTGCAATTCTTCCCGTTCCTTC[A/G]TGGATTTGAAGGCTCTCCTTTCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605778 MIM: 615811 | ||||||||||||||||||||
Literature Links: |
NIF3L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NIF3L1 - NGG1 interacting factor 3 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136039.2 | 309 | Missense Mutation | ATG,GTG | M,V 28 | NP_001129511.1 | |
NM_001142355.1 | 309 | Missense Mutation | ATG,GTG | M,V 1 | NP_001135827.1 | |
NM_001142356.1 | 309 | Missense Mutation | ATG,GTG | M,V 28 | NP_001135828.1 | |
NM_021824.3 | 309 | Missense Mutation | ATG,GTG | M,V 1 | NP_068596.2 | |
XM_005246742.3 | 309 | Missense Mutation | ATG,GTG | M,V 28 | XP_005246799.1 | |
XM_011511582.1 | 309 | Missense Mutation | ATG,GTG | M,V 28 | XP_011509884.1 | |
XM_011511583.1 | 309 | Missense Mutation | ATG,GTG | M,V 28 | XP_011509885.1 | |
XM_011511584.1 | 309 | Missense Mutation | ATG,GTG | M,V 28 | XP_011509886.1 | |
XM_011511585.1 | 309 | Missense Mutation | ATG,GTG | M,V 1 | XP_011509887.1 | |
XM_017004630.1 | 309 | Missense Mutation | ATG,GTG | M,V 28 | XP_016860119.1 |
PPIL3 - peptidylprolyl isomerase like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |