Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTGGTGTGTCTCCAGCCATCTA[C/T]TGATGGCCATTTTCAGAGTCCTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611397 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TANC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TANC1 - tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDSUB1 - WD repeat, sterile alpha motif and U-box domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128212.2 | 1368 | Missense Mutation | AAT,AGT | N,S 468 | NP_001121684.1 | |
NM_001128213.1 | 1368 | Missense Mutation | AAT,AGT | N,S 468 | NP_001121685.1 | |
NM_001307994.1 | 1368 | Missense Mutation | AAT,AGT | N,S 376 | NP_001294923.1 | |
NM_152528.2 | 1368 | Missense Mutation | AAT,AGT | N,S 468 | NP_689741.2 | |
XM_005246355.2 | 1368 | Missense Mutation | AAT,AGT | N,S 421 | XP_005246412.2 | |
XM_006712341.3 | 1368 | Missense Mutation | AAT,AGT | N,S 299 | XP_006712404.1 | |
XM_011510753.2 | 1368 | Missense Mutation | AAT,AGT | N,S 468 | XP_011509055.1 | |
XM_011510754.2 | 1368 | Missense Mutation | AAT,AGT | N,S 381 | XP_011509056.1 | |
XM_017003481.1 | 1368 | Intron | XP_016858970.1 | |||
XM_017003482.1 | 1368 | Missense Mutation | AAT,AGT | N,S 345 | XP_016858971.1 | |
XM_017003483.1 | 1368 | Missense Mutation | AAT,AGT | N,S 230 | XP_016858972.1 |